Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha Sarcoglycan Polyclonal Antibody
Alpha Sarcoglycan Polyclonal Antibody
Supplier: Thermo Scientific PA579990
Description
Alpha Sarcoglycan Polyclonal Antibody for Western Blot

Specifications
Alpha Sarcoglycan | |
Polyclonal | |
Unconjugated | |
SGCA | |
50 kDa dystrophin-associated glycoprotein; 50DAG; 50kD DAG; adhalin; ADL; Alpha-sarcoglycan; alpha-SG; Asg; DAG2; DMDA2; Dystroglycan 2; Dystroglycan2; dystroglycan-2; LGMD2D; limb girdle muscular dystrophy 2D; sarcoglycan alpha; sarcoglycan, alpha (50kDa dystrophin-associated glycoprotein); sarcoglycan, alpha (dystrophin-associated glycoprotein); SCARMD1; SG alpha; SG-alpha; Sgca | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20391, 303468, 6442 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
P82350, Q16586 | |
SGCA | |
E.coli-derived human SGCA recombinant protein (Position: Q24-H387). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction