Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Alpha Sarcoglycan Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537050
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537050 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537050 Supplier Invitrogen™ Supplier No. PA537050
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 50 kDa. Purity is >95% by SDS-PAGE.

The sarcoglycan transmembrane proteins are members of the dystrophin complex. Sarcoglycans cluster together to form a complex, which is localized in the cell membrane of skeletal, cardiac, and smooth muscle fibers. Four sarcoglycan subunit proteins, designated alpha-, beta-, gamma- and delta-sarcoglycan, form a complex on the skeletal muscle cell surface membrane. A genetic defect in any one of these proteins causes the loss or marked decrease of the whole sarcoglycan complex, which is observed in the autosomal recessive muscular dystrophy, sarcoglycanopathy. In smooth muscle, beta- and delta-sarcoglycans are associated with epsilon-sarcoglycan, a glycoprotein homologous to alpha-sarcoglycan. Additionally, a complete deficiency in delta-sarcoglycan is the cause of the Syrian hamster BIO.14 cardiomyopathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Alpha Sarcoglycan
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene SGCA
Gene Accession No. P82350, Q16586
Gene Alias 50 kDa dystrophin-associated glycoprotein; 50DAG; 50kD DAG; adhalin; ADL; Alpha-sarcoglycan; alpha-SG; Asg; DAG2; DMDA2; Dystroglycan 2; Dystroglycan2; dystroglycan-2; LGMD2D; limb girdle muscular dystrophy 2D; sarcoglycan alpha; sarcoglycan, alpha (50kDa dystrophin-associated glycoprotein); sarcoglycan, alpha (dystrophin-associated glycoprotein); SCARMD1; SG alpha; SG-alpha; Sgca
Gene Symbols SGCA
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 209-254 of human Sarcoglycan-alpha.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20391, 303468, 6442
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.