Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Alpha Sarcoglycan Recombinant Rabbit Monoclonal Antibody (3W3Q10)
GREENER_CHOICE

Catalog No. PIMA537982
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537982 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537982 Supplier Invitrogen™ Supplier No. MA537982
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: Rat heart, Rat skeletal muscle Immunogen sequence: VDALVTLLVP LLVALLLTLL LAYVMCCRRE GRLKRDLATS DIQMVHHCTI HGNTEELRQM AASREVPRPL STLPMFNVHT GERLPPRVDS AQVPLILDQH.

The sarcoglycan transmembrane proteins are members of the dystrophin complex. Sarcoglycans cluster together to form a complex, which is localized in the cell membrane of skeletal, cardiac, and smooth muscle fibers. Four sarcoglycan subunit proteins, designated alpha-, beta-, gamma- and delta-sarcoglycan, form a complex on the skeletal muscle cell surface membrane. A genetic defect in any one of these proteins causes the loss or marked decrease of the whole sarcoglycan complex, which is observed in the autosomal recessive muscular dystrophy, sarcoglycanopathy. In smooth muscle, beta- and delta-sarcoglycans are associated with epsilon-sarcoglycan, a glycoprotein homologous to alpha-sarcoglycan. Additionally, a complete deficiency in delta-sarcoglycan is the cause of the Syrian hamster BIO.14 cardiomyopathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Alpha Sarcoglycan
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 3W3Q10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3
Gene SGCA
Gene Accession No. P82350
Gene Alias 50 kDa dystrophin-associated glycoprotein; 50DAG; 50kD DAG; adhalin; ADL; Alpha-sarcoglycan; alpha-SG; Asg; DAG2; DMDA2; Dystroglycan 2; Dystroglycan2; dystroglycan-2; LGMD2D; limb girdle muscular dystrophy 2D; sarcoglycan alpha; sarcoglycan, alpha (50kDa dystrophin-associated glycoprotein); sarcoglycan, alpha (dystrophin-associated glycoprotein); SCARMD1; SG alpha; SG-alpha; Sgca
Gene Symbols SGCA
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20391, 303468
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.