Learn More
alpha-Smooth Muscle Actin Antibody (1A4 + ACTA2/791), Alexa Fluor™ 594, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | alpha-Smooth Muscle Actin |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Monoclonal |
| Clone | 1A4 + ACTA2/791 |
| Conjugate | Alexa Fluor 594 |
| Dilution | Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | AAT6, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTSA, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, sma, smooth muscle actin |
| Gene Symbols | ACTA2 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.