Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-Smooth Muscle Actin Mouse anti-Human, Clone: 132D3, Novus Biologicals™

Mouse Monoclonal Antibody
$340.50 - $499.50
Specifications
Antigen | alpha-Smooth Muscle Actin |
---|---|
Clone | 132D3 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence |
Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25942950
![]() |
Novus Biologicals
NBP25942950UL |
50 μL |
Each for $340.50
|
|
|||||
NBP259429X
![]() |
Novus Biologicals
NBP259429100UL |
100 μL |
Each for $499.50
|
|
|||||
Description
alpha-Smooth Muscle Actin Monoclonal specifically detects alpha-Smooth Muscle Actin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
alpha-Smooth Muscle Actin | |
1 mg/mL | |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Loading Controls, Stem Cell Markers | |
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
59 | |
Recombinant human ACTA2 (3-377aa) purified from E. coli | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
132D3 | |
Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence | |
Monoclonal | |
Purified | |
RUO | |
Human | |
AAT6, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, sma, smooth muscle actin | |
ACTA2 | |
IgG1 κ | |
Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title