Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-Synuclein Antibody (5C2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NBP104321
Description
alpha-Synuclein Monoclonal specifically detects alpha-Synuclein in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| alpha-Synuclein | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence | |
| Lewy body) 4, MGC110988, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, non-A4 component of amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, synuclein, alpha (non A4 component of amyloid precursor) | |
| Mouse | |
| 14.46 kDa | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience | |
| 6622 | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 5C2 | |
| Unconjugated | |
| P37840 | |
| SNCA | |
| This alpha-Synuclein Antibody (5C2) was developed against recombinant human alpha-synuclein (61-95) purified from E. coli (NP_0003360). | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human alpha-Synuclein (61-95 aa) mAb, clone 5C2, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human alpha-synuclein protein. This specifically recognizes alpha-synuclein only, particularly the NAC domain of alpha-synuclein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction