Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-Synuclein Antibody (OTI2F6), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271293AF405
Description
alpha-Synuclein Monoclonal specifically detects alpha-Synuclein in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
alpha-Synuclein | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SNCA | |
This alpha-Synuclein Antibody (OTI2F6) was developed against full length human recombinant protein of human alpha-Synuclein(NP_000336) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI2F6 | |
Western Blot, Immunohistochemistry | |
Lewy body) 4, MGC110988, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, non-A4 component of amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, synuclein, alpha (non A4 component of amyloid precursor) | |
Mouse | |
Affinity Purified | |
Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience | |
6622.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction