Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-Synuclein, Cys Glu134, p Ser129 Antibody (J18), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP318266
This item is not returnable.
View return policy
Description
alpha-Synuclein Monoclonal antibody specifically detects alpha-Synuclein in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Dot BlotSpecifications
| alpha-Synuclein | |
| Monoclonal | |
| 1 mg/ml | |
| Western Blot 1:500, Immunohistochemistry, Dot Blot | |
| alpha-synuclein, I+/--synuclein, MGC110988, NACP, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, PD1, SNCA, synuclein alpha-140, synuclein, alpha (non A4 component of amyloid precursor), truncated alpha synuclein | |
| Human alpha-Synuclein AA 124-134: AYEMP-pS-EEGYQ-Cys | |
| 100 μg | |
| Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience | |
| 6622 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Dot Blot | |
| J18 | |
| Unconjugated | |
| PBS pH 7.4, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction