Learn More
Invitrogen™ alpha Synuclein Monoclonal Antibody (3F8), FITC
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA545844
Description
A 1:1,000 dilution of MA5-45844 was sufficient for detection of Alpha Synuclein in 15 μg of human brain cell lysate by ECL immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Binds within the 101-140 aa region of human alpha synuclein, determined using WB and ELISA with corresponding peptides. Detects approximately 14 kDa band corresponding to Alpha synuclein.approximately Does not detect truncated alpha Synuclein (aa 1-114).
Alpha-synuclein is a member of the synuclein family, which also includes beta- and gamma-synuclein. These proteins are abundant in the brain and play a role in regulating synaptic transmission and membrane trafficking. Alpha- and beta-synuclein have been shown to selectively inhibit phospholipase D2, suggesting that they may also be involved in signaling pathways. Defects in the SNCA gene, which encodes alpha-synuclein, have been implicated in the pathogenesis of Parkinson's disease. Additionally, alpha-synuclein peptides are a major component of amyloid plaques in the brains of patients with Alzheimer's disease. Multiple alternatively spliced transcripts encoding different isoforms have been identified for this gene. Diseases associated with SNCA include Parkinson Disease 1, Autosomal Dominant and Dementia, Lewy Body.
Specifications
alpha Synuclein | |
Monoclonal | |
1 mg/mL | |
PBS, 9.1 mM sodium bicarbonate with 640.91 mM DMSO, 136.36 mM ethanolamine and no preservative; pH 7.4 | |
O55042, P37377, P37840 | |
SNCA | |
Mouse alpha synuclein aggregate. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Dot Blot, ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry | |
3F8 | |
FITC | |
SNCA | |
Alpha synuclein; alphaSYN; alpha-synuclein; NACP; non A-beta component of AD amyloid; non-A beta component of AD amyloid; non-A4 component of amyloid; Non-A4 component of amyloid precursor; PARK1; PARK4; PD1; Snca; Syn; synuclein alpha; synuclein alpha-140; synuclein, alpha; synuclein, alpha (non A4 component of amyloid precursor) | |
Mouse | |
Protein G | |
RUO | |
20617, 29219, 6622 | |
4°C, store in dark | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.