Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-synuclein Mouse anti-Human,Mouse, Unconjugated, Clone: Syn506, MilliporeSigma™

Catalog No. MABN25221MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN25221MI 100 μL
MABN25222MI 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN25221MI Supplier MilliporeSigma Supplier No. MABN2522100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alpha-synuclein Monoclonal antibody specifically detects alpha-synuclein in Human,Mouse samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-synuclein
Applications ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone Syn506
Conjugate Unconjugated
Gene Accession No. NP_000336.1
Gene Alias Non-A beta component of AD amyloid;Non-A4 component of amyloid precursor;NACP
Gene Symbols SNCA, NACP, PARK1
Host Species Mouse
Immunogen Recombinant human alpha-synuclein oxidized/nitrated in vitro with peroxynitrite.
Molecular Weight of Antigen 14.46 kDa calculated
Purification Method Protein G Purified
Quantity 100 μL
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone Syn506 is a mouse monoclonal antibody that detects misfolded alpha-synuclein. It targets an epitope within 20 amino acids from the N-terminal region.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.