Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-synuclein N103 fragment Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABN226025UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN2260-25UG 25 μg
ABN2260-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABN2260-25UG Supplier MilliporeSigma Supplier No. ABN226025UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-alpha-synuclein N103 fragment Antibody, Cat. No. ABN2260, is a highly specific rabbit polyclonal antibody that targets alpha-synuclein and has been tested for use in Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-synuclein N103 fragment
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in PBS with 0.02 mg/ml sodium azide and 40% glycerol.
Gene Alias Non-A beta component of AD amyloid;Non-A4 component of amyloid precursor;NACP
Gene Symbols SNCA;NACP;PARK1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to nine amino acids that precede the asparagine endopeptidase (AEP)-cleavage site at N103.
Molecular Weight of Antigen ∽38 kD observed (GFP Fusion protein); 14.46 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_000336
Test Specificity This rabbit polyclonal antibody detects human alpha-synuclein N103 fragment.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Product Type Antibodies
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.