Learn More
Invitrogen™ alpha Synuclein Polyclonal Antibody

Description
This antibody is predicted to react with bovine, porcine and non-human primate based on sequence homology.
Specifications
Specifications
| Antigen | alpha Synuclein |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | SNCA |
| Gene Accession No. | O55042, P37840 |
| Gene Alias | Alpha synuclein; alphaSYN; alpha-synuclein; NACP; non A-beta component of AD amyloid; non-A beta component of AD amyloid; non-A4 component of amyloid; Non-A4 component of amyloid precursor; PARK1; PARK4; PD1; Snca; Syn; synuclein alpha; synuclein alpha-140; synuclein, alpha; synuclein, alpha (non A4 component of amyloid precursor) |
| Gene Symbols | SNCA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.