Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alpha-Synuclein Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980607
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0607 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0607 Supplier Bioss Supplier No. BS0968R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Alpha-Synuclein belongs to the Synuclein family, which includes beta and gamma Synuclein, and is predominantly expressed in neurons, and concentrated at synaptic terminals. Synucleins may be involved in the regulation of dopamine release and transport and the modulation of synaptic vesicle function. Alpha-Synuclein is a small, presynaptic protein that is the major nonamyloid component of the pathological inclusions characteristic of a wide range of neurodegenerative disorders, collectively known as synucleinopathies. Alpha-Synuclein is involved in the formation of SNARE complexes, and most significantly, aggregated alpha-synuclein is one of the major components found in the Lewy bodies that occur in Parkinson′s disease (PD) and other neurodegenerative disorders. Mutation of the alpha-synuclein gene is associated with familial forms of PD. Alpha-Synuclein is known to reduce the fibrillization of the microtubule-associated protein, tau, to bind various targets such as 14-3-3, protein kinase C, synphilin-1, Elk and Tat-binding protein 1. The alpha and beta synucleins are found primarily in brain tissue where they are seen mainly in neuronal cytosol and presynaptic terminals. The antibody specific for alpha-synuclein does not cross react with beta-synuclein and will be useful for the confirmation of Parkinson′s disease and Lewy body dementia and other studies for the localization and detection of alpha-synuclein. Four alternatively spliced transcripts that encode two different isoforms of the alpha-synuclein protein have been identified.

Specifications

Antigen Alpha-Synuclein
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SNCA
Gene Accession No. P37840
Gene Alias Alpha synuclein; alphaSYN; Alpha-synuclein; MGC110988; MGC110988 antibody; NACP; NACP antibody; non A-beta component of AD amyloid; Non-A beta component of AD amyloid; non-A4 component of amyloid; Non-A4 component of amyloid precursor; OTTHUMP00000218549; OTTHUMP00000218551; OTTHUMP00000218552; OTTHUMP00000218553; OTTHUMP00000218554; PARK 1; PARK 1 antibody; PARK 4; PARK 4 antibody; PARK1; PARK1 antibody; PARK4; PARK4 antibody; PD 1 antibody; PD1; PD1 antibody; SNCA; SNCA antibody; Syn; synuclein alpha; synuclein alpha-140; synuclein, alpha; synuclein, alpha (non A4 component of amyloid precursor); SYUA
Gene Symbols SNCA
Host Species Rabbit
Immunogen Recombinant full length Alpha-Synuclein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6622
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.