Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alpha taxilin Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735700 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5700 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5700 Supplier Proteintech Group Inc Supplier No. 275581AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. NP220 (Nuclear protein 220), also designated Zinc finger protein 638 (ZNF638) or Cutaneous T-cell lymphoma-associated antigen se33-1, is a 1978 amino acid protein that contains one matrin-type zinc finger and two RRM (RNA recognition motif) domains, suggesting a role in transcriptional regulation. NP220 binds to double-stranded DNA fragments by recognizing clusters of cytidines. NP220 interacts with FHL-2 and is also thought to be phosphorylated by ATM or ATR upon DNA damage. It exists as five isoforms as a result of alternative splicing events. Isoform five of NP220 is a tumor-associated antigen found in several cutaneous T-cell lymphoma (CTCL), and in particular in mycosis fungoides patients and in Sezary syndrome patients.

Specifications

Antigen Alpha taxilin
Applications Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TXLNA
Gene Accession No. P40222
Gene Alias Alpha taxilin, IL14, RP4 622L5.4, taxilin alpha, TXLN, TXLNA
Gene Symbols TXLNA
Host Species Rabbit
Immunogen Alpha taxilin Fusion Protein Ag23001
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200081
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.