Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha TLR10 Mouse anti-Human, Unconjugated, Clone: 5C2C5, MilliporeSigma™

Catalog No. MABF20942MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF20942MI 25 μg
MABF20941MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF20942MI Supplier MilliporeSigma Supplier No. MABF209425UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alpha TLR10 Monoclonal antibody specifically detects alpha TLR10 in Human samples. It is validated for Flow Cytometry, Inhibits Activity/Function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha TLR10
Applications Flow Cytometry
Classification Monoclonal
Clone 5C2C5
Conjugate Unconjugated
Gene Accession No. NP_001017388
Gene Alias CD290
Gene Symbols TLR10, UNQ315/PRO358
Host Species Mouse
Immunogen Recombinant fragment corresponding to 455 amino acids from the N-terminal, extracellular domain of human Toll-like receptor 10.
Molecular Weight of Antigen 94.56 kDa calculated.
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone 5C2C5 is a mouse monoclonal antibody that detects human Toll-like receptor 10.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.