Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha TTP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589852
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589852 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589852 Supplier Invitrogen™ Supplier No. PA589852
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAEARSQPSA GPQLNALPDH SPLLQPGLAA LRRRAREAGV PLAPLPLTDS FLLRFLRARD FDLDLAWRLL KNYYKWRAEC PEISADLHPR SIIGLLKAGY HGVLRSRDPT; Positive Samples: Mouse liver, Mouse kidney, Rat liver; Cellular Location: Cytoplasm.

This gene encodes a soluble protein that binds alpha-trocopherol, a form of vitamin E, with high selectivity and affinity. This protein plays an important role in regulating vitamin E levels in the body by transporting vitamin E between membrane vesicles and facilitating the secretion of vitamin E from hepatocytes to circulating lipoproteins. Mutations in this gene cause hereditary vitamin E deficiency (ataxia with vitamin E deficiency, AVED) and retinitis pigmentosa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha TTP
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.81 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Ttpa
Gene Accession No. P41034, Q8BWP5
Gene Alias alpha tocopherol transfer protein; alpha TTP; alpha-tocopherol transfer protein; alphaTTP; alpha-TTP; ATTP; AVED; tocopherol (alpha) transfer protein; tocopherol (alpha) transfer protein (ataxia (Friedreich-like) with vitamin E deficiency); Tocopherol transfer protein alpha; TPP1; TTP; TTP1; TTPA
Gene Symbols Ttpa
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human TTPA (NP_0003611).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25571, 50500
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.