Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alpha TUBU Mouse anti-Human, Pig, Clone: TU-16, Abnova™

Catalog No. 89933554 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-554 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-554 Supplier Abnova Corporation Supplier No. MAB12938
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against pig Alpha Tubulin.

Specifications

Antigen Alpha TUBU
Applications ELISA, Immunocytochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone TU-16
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against pig Alpha Tubulin.
Dilution ELISA Immunocytochemistry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Immunoprecipitation (1-2 ug/mL) Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide).
Host Species Mouse
Immunogen Pig Alpha Tubulin.
Purification Method Caprylic acid and ammonium sulfate precipitation
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Pig brain microtubule protein MTP-1.
Target Species Human, Pig
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.