Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha Tubulin 3c Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281701AF750
Description
alpha Tubulin 3c Polyclonal antibody specifically detects alpha Tubulin 3c in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
alpha Tubulin 3c | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA | |
Alexa Fluor 750 | |
Alpha-tubulin 2, Alpha-tubulin 3C/D, bA408E5.3, TUBA2Tubulin alpha-2 chain, TUBA3D, tubulin alpha-3C/D chain, tubulin, alpha 2, tubulin, alpha 3c | |
Rabbit polyclonal alpha Tubulin 3c antibody was raised against a 16 amino acid peptide near the amino terminus of human alpha Tubulin 3c. The immunogen is located within amino acids 140 - 190 of alpha Tubulin 3c. Amino Acid Squence: MERLSVDYGKKSKLEF | |
0.1 mL | |
Signal Transduction | |
7278 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction