Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha Tubulin 3c Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281701DL594
This item is not returnable.
View return policy
Description
alpha Tubulin 3c Polyclonal antibody specifically detects alpha Tubulin 3c in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
alpha Tubulin 3c | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA | |
DyLight 594 | |
Alpha-tubulin 2, Alpha-tubulin 3C/D, bA408E5.3, TUBA2Tubulin alpha-2 chain, TUBA3D, tubulin alpha-3C/D chain, tubulin, alpha 2, tubulin, alpha 3c | |
Rabbit polyclonal alpha Tubulin 3c antibody was raised against a 16 amino acid peptide near the amino terminus of human alpha Tubulin 3c. The immunogen is located within amino acids 140 - 190 of alpha Tubulin 3c. Amino Acid Squence: MERLSVDYGKKSKLEF | |
0.1 mL | |
Signal Transduction | |
7278 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction