Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha Tubulin 4a Antibody (B-5-1-2), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB12011304AF594
Description
alpha Tubulin 4a Monoclonal specifically detects alpha Tubulin 4a in Human, Mouse, Rat, Bovine, Chicken, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
alpha Tubulin 4a | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
TUBA4A | |
Sarkosyl-resistant filaments from Stronglycentrotus purpuratus (sea urchin). | |
0.1 mL | |
Primary | |
Recognizes an epitope located at the C-terminal end of the alpha-tubulin isoform in a variety of organisms. | |
Store at 4°C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
B-5-1-2 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
alpha 1 (testis specific), Alpha-tubulin 1, FLJ30169, H2-ALPHA, Testis-specific alpha-tubulin, TUBA1, Tubulin alpha-1 chain, tubulin alpha-4A chain, Tubulin H2-alpha, tubulin, alpha 1, tubulin, alpha 4a | |
Mouse | |
Protein A or G purified | |
Cell Biology, Cytoskeleton Markers, Stem Cell Markers | |
7277.0 | |
Human, Mouse, Rat, Bovine, Chicken, Primate | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction