Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha Tubulin 4a Antibody (B-5-1-2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB12011304
Description
alpha Tubulin 4a Monoclonal specifically detects alpha Tubulin 4a in Human, Mouse, Rat, Bovine, Chicken, Primate samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Microarray.Specifications
| alpha Tubulin 4a | |
| Monoclonal | |
| 2.0 mg/ml | |
| Western Blot 0.25-0.5 ug/ml, Simple Western, Immunocytochemistry/Immunofluorescence 0.5-1 ug/ml, Immunoprecipitation 1:10-1:500, Microarray | |
| TUBA4A | |
| Sarkosyl-resistant filaments from Stronglycentrotus purpuratus (sea urchin). | |
| Affinity Purified | |
| RUO | |
| 7277 | |
| Human, Mouse, Rat, Bovine, Chicken, Primate | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoassay | |
| B-5-1-2 | |
| Unconjugated | |
| alpha 1 (testis specific), Alpha-tubulin 1, FLJ30169, H2-ALPHA, Testis-specific alpha-tubulin, TUBA1, Tubulin alpha-1 chain, tubulin alpha-4A chain, Tubulin H2-alpha, tubulin, alpha 1, tubulin, alpha 4a | |
| Mouse | |
| ∼50 kDa | |
| 0.1 mL | |
| Primary | |
| Recognizes an epitope located at the C-terminal end of the alpha-tubulin isoform in a variety of organisms. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction