Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha Tubulin 4a Antibody (B-5-1-2), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB12011304IR
Description
alpha Tubulin 4a Monoclonal specifically detects alpha Tubulin 4a in Human, Mouse, Rat, Bovine, Chicken, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
alpha Tubulin 4a | |
Monoclonal | |
DyLight 755 | |
alpha 1 (testis specific), Alpha-tubulin 1, FLJ30169, H2-ALPHA, Testis-specific alpha-tubulin, TUBA1, Tubulin alpha-1 chain, tubulin alpha-4A chain, Tubulin H2-alpha, tubulin, alpha 1, tubulin, alpha 4a | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Recognizes an epitope located at the C-terminal end of the alpha-tubulin isoform in a variety of organisms. | |
Store at 4C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
B-5-1-2 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
TUBA4A | |
Sarkosyl-resistant filaments from Stronglycentrotus purpuratus (sea urchin). | |
0.1 mL | |
Cell Biology, Cytoskeleton Markers, Stem Cell Markers | |
7277 | |
Human, Mouse, Rat, Bovine, Chicken, Primate | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction