Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha Tubulin Antibody (961258), Alexa Fluor™ 750, R&D Systems™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB93441AF750
Description
alpha Tubulin Monoclonal specifically detects alpha Tubulin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry.Specifications
alpha Tubulin | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
TUBA1A | |
This alpha Tubulin Antibody (961258) was developed against alpha Tubulin. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
961258 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry | |
Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA3, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific | |
Mouse | |
Protein A or G purified | |
Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Loading Controls, Neuroscience | |
7846.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction