Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha Tubulin Antibody (DM1A), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100690APC
Description
alpha Tubulin Monoclonal specifically detects alpha Tubulin in Human, Mouse, Rat, Porcine, Avian, Bovine, Canine, Chicken, Chinese Hamster, Drosophila, Fungi, Guinea Pig, Hamster, Parasite, Primate, Monkey, Rabbit, Xenopus, Yeast samples. It is validated for Flow Cytometry.Specifications
alpha Tubulin | |
Monoclonal | |
APC | |
Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA3, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific | |
Mouse | |
50 kDa | |
0.1 mL | |
Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Loading Controls, Neuroscience | |
7846 | |
Human, Mouse, Rat, Pig, Avian, Bovine, Canine, Chicken, Chinese Hamster, Drosophila, Fungi, Guinea Pig, Hamster, Parasite, Primate, Monkey, Rabbit, Xenopus species, Yeast | |
Purified |
Flow Cytometry | |
DM1A | |
Flow Cytometry | |
TUBA1A | |
This alpha Tubulin Antibody (DM1A) was developed against native chicken brain microtubules. | |
Protein G purified | |
RUO | |
Primary | |
This alpha Tubulin Antibody (DM1A) does not cross-react with beta Tubulin. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction