Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha Tubulin Antibody (DM1A), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP323258
Description
alpha Tubulin Monoclonal antibody specifically detects alpha Tubulin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
alpha Tubulin | |
Monoclonal | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
DM1A | |
Western Blot 1:500, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation | |
Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA3, tublin, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific | |
Clone DM1A was generated from purified chick brain tubulin. The antibody recognizes an epitope within the C-terminal end of alpha Tubulin isoforms. The epitope is highly conserved in alpha Tubulin isoforms from most vertebrate species. [UniProt# P02552] | |
0.1 mL | |
Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Loading Controls, Neuroscience | |
7846 | |
Store at -20°C. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction