Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha Tubulin Antibody (YOL1/34) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP280569
Description
alpha Tubulin Monoclonal specifically detects alpha Tubulin in Human, Mouse, Rat, Drosophila, Fungi, Yeast samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Radioimmunoassay, Flow (Intracellular).Specifications
| alpha Tubulin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot : 1:100-1:2000, Flow Cytometry, ELISA : 10 μg/mL, Immunohistochemistry : 1:10-1:500, Immunocytochemistry/Immunofluorescence : 1:10-1:500, Immunohistochemistry-Frozen : 1:10-1:500, Radioimmunoassay 1:100-1:2000, Flow (Intracellular) | |
| Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA3, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific | |
| Rat | |
| 50 kDa | |
| 0.25 mg | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Loading Controls, Neuroscience | |
| 7846 | |
| Human, Mouse, Rat, Drosophila, Fungi, Yeast | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Radioimmune Assays (RIA) | |
| YOL1/34 | |
| Unconjugated | |
| PBS | |
| TUBA1A | |
| This alpha Tubulin Antibody (YOL1/34) - Azide and BSA Free was developed against yeast tubulin. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Cross reacts with Arabidopsis, Saccharomyces, Platyzoma, Ashbya, and Naegleria. Based on sequence similarity, is expected to react with: Birds and Mammals. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction