Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha Tubulin Antibody (YOL1/34), DyLight 755, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB1001639IR
Description
alpha Tubulin Monoclonal specifically detects alpha Tubulin in Human, Mouse, Rat, Drosophila, Fungi, Yeast samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Flow (Intracellular).Specifications
alpha Tubulin | |
Monoclonal | |
DyLight 755 | |
Q71U36 | |
TUBA1A | |
This alpha Tubulin Antibody (YOL1/34) was made from yeast tubulin. | |
0.25 mL | |
Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Loading Controls, Neuroscience | |
7846 | |
Human, Mouse, Rat, Drosophila, Fungi, Yeast | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
YOL1/34 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Flow (Intracellular) | |
Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA3, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
This alpha Tubulin Antibody (YOL1/34) detects an epitope on alpha tubulin between the amino acids 414-422. It has a higher affinity for fixed microtubules (formaldehyde or glutaraldehyde) than native microtubules when performing IHC. | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction