Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha Tubulin Monoclonal Antibody (B-5-1-2)
GREENER_CHOICE

Catalog No. 322500
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-250-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-250-0 Supplier Invitrogen™ Supplier No. 322500
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

alpha Tubulin Monoclonal Antibody for ICC/IF, Western Blot

Alpha Tubulin is a part of a heterodimer consisting of alpha and beta tubulin subunits. The alpha and beta tubulins, which are each about 55 kDa MW, are homologous but not identical. Alpha-beta tubulin heterodimer is the basic building block of microtubules, and this intracellular cylindrical filamentous structure is present in almost eukaryotic cells. Microtubules serve as structural supports and lines of transport within the cell, as well as serving a key role in mitosis. Microtubules of the eukaryotic cytoskeleton perform essential and diverse functions and are composed of a heterodimer of alpha and beta tubulins. The alpha and beta tubulins represent the major components of microtubules, while gamma tubulin plays a critical role in the nucleation of microtubule assembly. There are multiple alpha and beta tubulin genes, which are highly conserved among species. The gene that encodes alpha tubulin is highly similar to the mouse and rat Tuba1 gene. Northern blotting studies have shown that the gene expression is predominantly found in morphologically differentiated neurologic cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha Tubulin
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone B-5-1-2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene TUBAL3
Gene Accession No. A6NHL2, P05213, P0DPH7, P68363, P68366, P68368, P68369, P68370, P68373, Q3KRE8, Q3UX10, Q5XIF6, Q6AYZ1, Q6P9V9, Q6PEY2, Q71U36, Q9BQE3, Q9BVA1, Q9CWF2
Gene Alias 1t; 4 tubulin; a1t; ALPHA 84C; alpha tubulin; alpha tubulin 84B; alpha[[1]] tubulin; alpha[[1]]-tubulin; alpha1; alpha1 tubulin; alpha-1 tubulin; alpha1t; alpha1tub; alpha1tub84B; alpha1-tubulin; alpha-2; alpha84B; alphaT; alphat-1; alphatub; alpha-tub; alphaTub1; alphatub84; alphaTUB84B; alpha-tub84B; alphaTub84B-PA; alphatubulin; alpha-tubulin; Alpha-tubulin 1; Alpha-tubulin 2; Alpha-tubulin 3; Alpha-tubulin 3C; alpha-tubulin 3C/D; Alpha-tubulin 3D; Alpha-tubulin 3E; alpha-tubulin 4; Alpha-tubulin 6; alphaTubulin 84B; alpha-tubulin 84B; alpha-Tubulin at 84B; alpha-tubulin I; Alpha-tubulin II; alpha-tubulin III; Alpha-tubulin isotype M-alpha-1; alpha-tubulin isotype M-alpha-2; Alpha-tubulin isotype M-alpha-4; alpha-tubulin isotype M-alpha-6; alpha-tubulin TUB1; alpha-tubulin ubiquitous; alphaTubulin84B; Alpha-Tubulin84B; ALS22; anon-EST:Liang-1.59; anon-EST:Liang-2.30; AT25469p; aTub84B; a-tub84B; bA408E5.3; B-ALPHA-1; bcm948; BEST:LD32507; cb944; CG1913; CG1913-PA; chr3R:2914276..2914416; clone 1.59; clone 2.30; Detyrosinated tubulin alpha-1A chain; Detyrosinated tubulin alpha-1B chain; Detyrosinated tubulin alpha-1C chain; Detyrosinated tubulin alpha-3 chain; Detyrosinated tubulin alpha-3C chain; Detyrosinated tubulin alpha-3D chain; Detyrosinated tubulin alpha-3E chain; DM1alpha; Dmel\CG1913; Dmel_CG1913; DTA1; fb22g06; FLJ30169; H2 ALPHA; H2-ALPHA; hum-a-tub1; hum-a-tub2; I79_019110; K-ALPHA-1; l(3)84Bd; l(3)g3; lis3; LOC100726777; LOC100731885; LOC100857858; LOC101118307; LOC106557476; M[a]4; M[a]6; ms(3)nc33; nc33; RGD1565476; similar to alpha-tubulin isoform 1; similar to tubulin alpha 1; T; Talpha1; Talpha1 alpha-tubulin; TBA1_DROME; Testis-specific alpha-tubulin; TUA; TUA1; TUA2; TUA3; Tub; Tub 84B; TUB1; Tub1C; Tub84B; TUBA1; Tuba-1; tuba1 protein; TUBA1/3/4; Tuba1a; tuba1a.L; tuba1a-a; tuba1a-b; TUBA1B; TUBA1C; tuba1l; tuba1l2; TUBA2; TUBA3; TUBA3C; TUBA3D; TUBA3E; Tuba4; Tuba4a; TUBA5; Tuba6; tuba8.S; tubA84B; tubal2; Tubalpha1; tubulin; tubulin 1 alpha-chain; tubulin 1alpha; tubulin alpha; tubulin alpha 1a; tubulin alpha 1a L homeolog; tubulin alpha 1b; tubulin alpha 1c; tubulin alpha 3; tubulin alpha 3c; tubulin alpha 3e; tubulin alpha 4a; tubulin alpha 6; tubulin alpha 8 S homeolog; tubulin alpha1; tubulin alpha-1; tubulin alpha-1 chain; tubulin alpha1 promoter; Tubulin alpha-1A chain; tubulin alpha-1B chain; Tubulin alpha-1C chain; tubulin alpha-1C chain-like; tubulin alpha-2 chain; tubulin alpha-3; Tubulin alpha-3 chain; Tubulin alpha-3C chain; tubulin alpha-3C/D chain; Tubulin alpha-3D chain; Tubulin alpha-3E chain; tubulin alpha-4 chain; Tubulin alpha-4A chain; Tubulin alpha-5 chain; tubulin alpha-5 chain-like; tubulin alpha-6 chain; tubulin alpha-8 chain; tubulin alpha-ubiquitous chain; tubulin B-alpha-1; Tubulin H2-alpha; tubulin K-alpha-1; tubulin(alpha1); tubulin, alpha 1; tubulin, alpha 1 (testis specific); tubulin, alpha 1, like; tubulin, alpha 1, like 2; tubulin, alpha 1a; tubulin, alpha 1b; tubulin, alpha 1c; tubulin, alpha 2; tubulin, alpha 3c; tubulin, alpha 3e; tubulin, alpha 4; tubulin, alpha 4a; tubulin, alpha 5; tubulin, alpha 6; tubulin, alpha, brain-specific; tubulin, alpha, ubiquitous; tubulin1alpha; tubulin84B; Tubulin-alpha1; Unknown (protein for MGC:133894); unnamed protein product; wu:fb22g06; wu:fb37a10; XELAEV_18019849mg; XELAEV_18020901mg; YML085C
Gene Symbols LOC106557476, Tuba1a, Tuba1b, TUBA1C, TUBA3C, TUBA3E, TUBA4A, TUBAL3, TUBB2B
Host Species Mouse
Immunogen Sarkosyl-resistant filament from sea urchin sperm axonemes.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10376, 106557476, 112714, 22142, 22143, 22145, 22146, 238463, 291081, 291287, 300218, 316531, 347733, 478702, 478918, 487148, 500929, 608138, 608864, 64158, 7277, 7278, 73710, 7846, 79861, 84790
Target Species Human, Mouse, Rat, Canine, Monkey, Sea urchin
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.