Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alpha Tubulin Mouse anti-Bovine, Human, Mouse, Clone: 236-10501, Abnova™

Catalog No. 89035722 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-722 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-722 Supplier Abnova Corporation Supplier No. MAB13986

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against native bovine alpha-tubulin.

  • High specificity
  • High reactivity

Specifications

Antigen Alpha Tubulin
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 236-10501
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native bovine alpha-tubulin.
Dilution Flow Cytometry (1:100 - 1:500) Immunocytochemistry (1:100 - 1:500) Immunofluorescence (1:100 - 1:500) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:20 - 1:200) Western Blot (1:1000 - 1:5000) The optimal working dil
Formulation Lyophilized from PBS.
Host Species Mouse
Immunogen Native purified bovine alpha-tubulin.
Purification Method Biochemical fractionation
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bovine, Human, Mouse
Content And Storage Store at -20°C.
Reconstitute in PBS, containing 1% BSA and 2 mM sodium azide.
Reconstituted solution should remain stable for 6 months when stored at 4°C. For longer storage, stored the reconstituted solution at -20°C.
Aliquot to avo
Form Lyophilized
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.