Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Tubulin Mouse anti-Human, Mouse, Rat, HRP, Clone: 1E4C11, Proteintech
SDP

Catalog No. 50553248 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-553-248 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-553-248 Supplier Proteintech Group Inc Supplier No. HRP66031100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Alpha Tubulin is a part of a heterodimer consisting of alpha and beta tubulin subunits. The alpha and beta tubulins, which are each about 55 kDa MW, are homologous but not identical. Alpha-beta tubulin heterodimer is the basic building block of microtubules, and this intracellular cylindrical filamentous structure is present in almost eukaryotic cells. Microtubules serve as structural supports and lines of transport within the cell, as well as serving a key role in mitosis. Microtubules of the eukaryotic cytoskeleton perform essential and diverse functions and are composed of a heterodimer of alpha and beta tubulins. The alpha and beta tubulins represent the major components of microtubules, while gamma tubulin plays a critical role in the nucleation of microtubule assembly. There are multiple alpha and beta tubulin genes, which are highly conserved among species. The gene that encodes alpha tubulin is highly similar to the mouse and rat Tuba1 gene. Northern blotting studies have shown that the gene expression is predomitly found in morphologically differentiated neurologic cells.

Specifications

Antigen alpha Tubulin
Applications Western Blot
Classification Monoclonal
Clone 1E4C11
Concentration 1.33 mg/mL
Conjugate HRP
Formulation PBS with 50% glycerol and 0.02% thimerosal; pH 7.3
Gene TUBA1B
Gene Accession No. P05213, P68363, Q6P9V9
Gene Alias Alpha tubulin ubiquitous, alpha tubulin,a tubulin, HRP Tubulin Alpha, K ALPHA 1, TUBA1B, tubulin, Tubulin Alpha, Tubulin alpha 1B chain, Tubulin alpha ubiquitous chain, Tubulin K alpha 1, tubulin, alpha 1b
Gene Symbols Tuba1b
Host Species Mouse
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10376, 22143, 500929
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.