Learn More
Invitrogen™ TUBA3C Polyclonal Antibody

Description
Sequence homology: Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Specifications
Specifications
| Antigen | TUBA3C |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | TUBA3C |
| Gene Accession No. | P0DPH7 |
| Gene Alias | alpha tubulin 3C; alpha-tubulin 2; Alpha-tubulin 3C; alpha-tubulin 3C/D; Alpha-tubulin 3D; bA408E5.3; Detyrosinated tubulin alpha-3C chain; Detyrosinated tubulin alpha-3D chain; TUBA2; TUBA3C; TUBA3D; tubulin alpha 3c; tubulin alpha-2 chain; Tubulin alpha-3C chain; tubulin alpha-3C/D chain; Tubulin alpha-3D chain; tubulin, alpha 2; tubulin, alpha 3c |
| Gene Symbols | TUBA3C |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.