Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TUBA1C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598220
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598220 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598220 Supplier Invitrogen™ Supplier No. PA598220
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TUBA1C Polyclonal Antibody for Western Blot, IHC (P), IP, ELISA

TUBA1C (Tubulin Alpha 1c) is a Protein Coding gene. Diseases associated with TUBA1C include Distal Hereditary Motor Neuropathy, Type Ii. Among its related pathways are Development Slit-Robo signaling and Gap junction. Gene Ontology (GO) annotations related to this gene include GTP binding and structural molecule activity. An important paralog of this gene is TUBA1B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUBA1C
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TUBA1C
Gene Accession No. P68373, Q9BQE3
Gene Alias alpha-tubulin 6; Alpha-tubulin isotype M-alpha-6; bcm948; Detyrosinated tubulin alpha-1C chain; M[a]6; Tuba1c; TUBA6; tubulin alpha 1c; tubulin alpha-1C chain; Tubulin alpha-6 chain; tubulin, alpha 1C; tubulin, alpha 6
Gene Symbols TUBA1C
Host Species Rabbit
Immunogen Recombinant Human Tubulin alpha-1C chain protein (29-184AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22146, 84790
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.