Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUBA3C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598436
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598436 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598436 Supplier Thermo Scientific Supplier No. PA598436
Add to Cart
Add to Cart

TUBA3C Polyclonal Antibody

TUBA3C Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen TUBA3C
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TUBA3C
Gene Accession No. P0DPH7
Gene Alias alpha tubulin 3C; alpha-tubulin 2; Alpha-tubulin 3C; alpha-tubulin 3C/D; Alpha-tubulin 3D; bA408E5.3; Detyrosinated tubulin alpha-3C chain; Detyrosinated tubulin alpha-3D chain; TUBA2; TUBA3C; TUBA3D; tubulin alpha 3c; tubulin alpha-2 chain; Tubulin alpha-3C chain; tubulin alpha-3C/D chain; Tubulin alpha-3D chain; tubulin, alpha 2; tubulin, alpha 3c
Gene Symbols TUBA3C
Host Species Rabbit
Immunogen Recombinant Human Tubulin alpha-3C/D chain protein (161-418AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7278
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.