Learn More
Invitrogen™ TUBA4A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113679
Description
Immunogen sequence: YAKRAFVHWY VGEGMEEGEF SEAREDMAAL EKDYEEVGID SYEDEDEGEE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%.
Alpha Tubulin is a part of a heterodimer consisting of alpha and beta tubulin subunits. The alpha and beta tubulins, which are each about 55 kDa, are homologous but not identical. The alpha-beta tubulin heterodimer is the basic building block of microtubules, and this intracellular cylindrical filamentous structure is present in almost all eukaryotic cells. Microtubules serve as structural supports and lines of transport within the cell, as well as serving a key role in mitosis. Microtubules of the eukaryotic cytoskeleton perform essential and diverse functions and are composed of a heterodimer of alpha and beta tubulins. The alpha and beta tubulins represent the major components of microtubules, while gamma tubulin plays a critical role in the nucleation of microtubule assembly. There are multiple alpha and beta tubulin genes, which are highly conserved among species. The gene that encodes alpha tubulin is highly similar to the mouse and rat Tuba1 gene. Northern blotting studies have shown that the gene expression is predominantly found in morphologically differentiated neurologic cells.
Specifications
| TUBA4A | |
| Polyclonal | |
| Unconjugated | |
| TUBA4A | |
| Alpha-tubulin 1; Alpha-tubulin 4; alpha-tubulin isotype M-alpha-4; ALS22; H2-ALPHA; M[a]4; Testis-specific alpha-tubulin; TUBA1; Tuba4; TUBA4A; Tubulin alpha; tubulin alpha 4a; tubulin alpha-1 chain; Tubulin alpha-4 chain; tubulin alpha-4A chain; Tubulin H2-alpha; tubulin, alpha 1 (testis specific); tubulin, alpha 4; tubulin, alpha 4a | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22145, 7277 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P68366, P68368 | |
| TUBA4A | |
| Synthetic peptide directed towards the middle region of human TUBA4A. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.