Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TUBA1C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107044
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107044 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107044 Supplier Invitrogen™ Supplier No. PA5107044
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Dog: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%; Zebrafish: 100%.

TUBA1C (Tubulin Alpha 1c) is a Protein Coding gene. Diseases associated with TUBA1C include Distal Hereditary Motor Neuropathy, Type Ii. Among its related pathways are Development Slit-Robo signaling and Gap junction. Gene Ontology (GO) annotations related to this gene include GTP binding and structural molecule activity. An important paralog of this gene is TUBA1B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUBA1C
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene TUBA1C
Gene Accession No. P68373, Q9BQE3
Gene Alias alpha-tubulin 6; Alpha-tubulin isotype M-alpha-6; bcm948; Detyrosinated tubulin alpha-1C chain; M[a]6; Tuba1c; TUBA6; tubulin alpha 1c; tubulin alpha-1C chain; Tubulin alpha-6 chain; tubulin, alpha 1C; tubulin, alpha 6
Gene Symbols TUBA1C
Host Species Rabbit
Immunogen Synthetic peptide located within the following region: EAWARLDHKF DLMYAKRAFV HWYVGEGMEE GEFSEAREDM AALEKDYEEV (aa 386-435).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22146, 84790
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.