Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alpha-Tubulin Rabbit, Clone: RM113, Abnova™

Catalog No. 89933555 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-555 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-555 Supplier Abnova Corporation Supplier No. MAB12827
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against synthetic peptide of human alpha-Tubulin.

Microtubules of the eukaryotic cytoskeleton perform essential and diverse functions and are composed of a heterodimer of alpha and beta tubulins. The genes encoding these microtubule constituents belong to the tubulin superfamily, which is composed of six distinct families. Genes from the alpha, beta and gamma tubulin families are found in all eukaryotes. The alpha and beta tubulins represent the major components of microtubules, while gamma tubulin plays a critical role in the nucleation of microtubule assembly. There are multiple alpha and beta tubulin genes, which are highly conserved among species. This gene encodes alpha tubulin and is highly similar to mouse and rat Tuba1 gene. Northern blotting studies have shown that the gene expression is predominantly found in morphologically differentiated neurologic cells. This gene is one of three alpha-tubulin genes in a cluster on chromosome 12q. [provided by RefSeq]

Specifications

Antigen Alpha-Tubulin
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone RM113
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of human alpha-Tubulin.
Dilution ELISA Flow Cytometry Immunocytochemistry (1:200) Immunohistochemistry (1:200) Immunoprecipitation (1:200) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Gene Alias B-ALPHA-1/FLJ25113/LIS3/TUBA3
Gene Symbols TUBA1A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus region of human alpha-tubulin.
Purification Method Protein A affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7846/10376
Test Specificity This antibody reacts to alpha-Tubulin, including Tubulin alpha-1A chain and Tubulin alpha-1B chain.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.