Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha Tubulin Rat anti-Human, Mouse, Rat, Drosophila, Fungi, Yeast, FITC, Clone: YOL1/34, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB1001639F0.25ML
Description
alpha Tubulin Monoclonal specifically detects alpha Tubulin in Human, Mouse, Rat, Drosophila, Fungi, Yeast samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Flow (Intracellular).Specifications
alpha Tubulin | |
Monoclonal | |
FITC | |
Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA3, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific | |
This alpha Tubulin Antibody (YOL1/34) was made from yeast tubulin. | |
0.25 mL | |
Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Loading Controls, Neuroscience | |
7846 | |
Store at 4°C in the dark. | |
IgG2a |
ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
YOL1/34 | |
PBS | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Drosophila, Fungi, Yeast | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction