Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ alpha Tubulin Recombinant Protein Antigen
SDP

Catalog No. NBP254687X Shop All R&D Systems Products
Change view
Click to view available options
Protein Length:
ATYAPVISAEKAYHEQLSVAEITN
LEHGIQPDGQMPSDKTIGGGDDSFNTFF
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Protein Length
NBP254687X LEHGIQPDGQMPSDKTIGGGDDSFNTFF
NBP254708X ATYAPVISAEKAYHEQLSVAEITN
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP254687X Supplier Novus Biologicals™ Supplier No. NBP254687PEP
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human alpha Tubulin The alpha Tubulin Recombinant Protein Antigen is derived from E. coli. The alpha Tubulin Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

Specifications

Protein Length LEHGIQPDGQMPSDKTIGGGDDSFNTFF
Purity >80% by SDS-PAGE and Coomassie blue staining
Common Name TUBA1A Recombinant Protein Antigen
Content And Storage Store at −20°C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4
For Use With (Application) AC
Gene Alias Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA3, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific
Gene Symbol TUBA1A
Label Type Unlabeled
Molecular Weight (g/mol) 20 kDa
Product Type Recombinant Protein Antigen
Quantity 0.1mL
Regulatory Status RUO
Research Category Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Loading Controls, Neuroscience
Source E.coli
Specific Reactivity Human
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.