Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha Tubulin Recombinant Rabbit Monoclonal Antibody (RM113), Biotin
GREENER_CHOICE

Catalog No. PIMA527888
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527888 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527888 Supplier Invitrogen™ Supplier No. MA527888
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Alpha Tubulin is a part of a heterodimer consisting of alpha and beta tubulin subunits. The alpha and beta tubulins, which are each about 55 kDa MW, are homologous but not identical. Alpha-beta tubulin heterodimer is the basic building block of microtubules, and this intracellular cylindrical filamentous structure is present in almost eukaryotic cells. Microtubules serve as structural supports and lines of transport within the cell, as well as serving a key role in mitosis. Microtubules of the eukaryotic cytoskeleton perform essential and diverse functions and are composed of a heterodimer of alpha and beta tubulins. The alpha and beta tubulins represent the major components of microtubules, while gamma tubulin plays a critical role in the nucleation of microtubule assembly. There are multiple alpha and beta tubulin genes, which are highly conserved among species. The gene that encodes alpha tubulin is highly similar to the mouse and rat Tuba1 gene. Northern blotting studies have shown that the gene expression is predominantly found in morphologically differentiated neurologic cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha Tubulin
Applications ChIP Assay, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone RM113
Concentration 0.1 mg/mL
Conjugate Biotin
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
Gene TUBAL3
Gene Accession No. A6NHL2, P05213, P0DPH7, P68363, P68366, P68368, P68369, P68370, P68373, Q3KRE8, Q3UX10, Q5XIF6, Q6AYZ1, Q6P9V9, Q6PEY2, Q71U36, Q9BQE3, Q9BVA1, Q9CWF2
Gene Alias alpha-tubulin 84B; 1t; a1t; ALPHA 84C; alpha tubulin; alpha tubulin 84B; alpha[[1]] tubulin; alpha[[1]]-tubulin; alpha1; alpha1 tubulin; alpha1t; alpha1tub; alpha1tub84B; alpha1-tubulin; alpha84B; alphaT; alphat-1; alphatub; alpha-tub; alphaTub1; alphatub84; alphaTUB84B; alpha-tub84B; alphaTub84B-PA; alphatubulin; alpha-tubulin; Alpha-tubulin 1; Alpha-tubulin 2; Alpha-tubulin 3; Alpha-tubulin 3C; alpha-tubulin 3C/D; Alpha-tubulin 3D; Alpha-tubulin 3E; alpha-tubulin 4; Alpha-tubulin 6; alphaTubulin 84B; alpha-Tubulin 84B; alpha-Tubulin at 84B; alpha-tubulin I; Alpha-tubulin II; alpha-tubulin III; Alpha-tubulin isotype M-alpha-1; alpha-tubulin isotype M-alpha-2; Alpha-tubulin isotype M-alpha-4; alpha-tubulin isotype M-alpha-6; alpha-tubulin TUB1; alpha-tubulin ubiquitous; alphaTubulin84B; Alpha-Tubulin84B; ALS22; anon-EST:Liang-1.59; anon-EST:Liang-2.30; AT25469p; aTub84B; a-tub84B; bA408E5.3; B-ALPHA-1; bcm948; BEST:LD32507; cb944; CG1913; CG1913-PA; chr3R:2914276..2914416; clone 1.59; clone 2.30; Detyrosinated tubulin alpha-1A chain; Detyrosinated tubulin alpha-1B chain; Detyrosinated tubulin alpha-1C chain; Detyrosinated tubulin alpha-3 chain; Detyrosinated tubulin alpha-3C chain; Detyrosinated tubulin alpha-3D chain; Detyrosinated tubulin alpha-3E chain; DM1alpha; Dmel\CG1913; Dmel_CG1913; DTA1; fb22g06; FLJ30169; H2 ALPHA; H2-ALPHA; hum-a-tub1; hum-a-tub2; I79_019110; K-ALPHA-1; l(3)84Bd; l(3)g3; lis3; LOC100726777; LOC100731885; LOC100857858; LOC101118307; LOC106557476; M[a]4; M[a]6; ms(3)nc33; nc33; RGD1565476; similar to alpha-tubulin isoform 1; similar to tubulin alpha 1; T; Talpha1; Talpha1 alpha-tubulin; TBA1_DROME; Testis-specific alpha-tubulin; Tub; Tub 84B; TUB1; Tub1C; Tub84B; TUBA1; Tuba-1; tuba1 protein; TUBA1/3/4; Tuba1a; tuba1a.L; tuba1a-a; tuba1a-b; TUBA1B; TUBA1C; tuba1l; tuba1l2; TUBA2; TUBA3; TUBA3C; TUBA3D; TUBA3E; Tuba4; Tuba4a; TUBA5; Tuba6; tuba8.S; tubA84B; tubal2; Tubalpha1; tubulin; tubulin 1 alpha-chain; tubulin 1alpha; tubulin alpha; tubulin alpha 1a; tubulin alpha 1a L homeolog; tubulin alpha 1b; tubulin alpha 1c; tubulin alpha 3; tubulin alpha 3c; tubulin alpha 3e; tubulin alpha 4a; tubulin alpha 6; tubulin alpha 8 S homeolog; tubulin alpha1; tubulin alpha-1 chain; tubulin alpha1 promoter; Tubulin alpha-1A chain; tubulin alpha-1B chain; Tubulin alpha-1C chain; tubulin alpha-1C chain-like; tubulin alpha-2 chain; Tubulin alpha-3 chain; Tubulin alpha-3C chain; tubulin alpha-3C/D chain; Tubulin alpha-3D chain; Tubulin alpha-3E chain; tubulin alpha-4 chain; Tubulin alpha-4A chain; Tubulin alpha-5 chain; tubulin alpha-5 chain-like; tubulin alpha-6 chain; tubulin alpha-8 chain; tubulin alpha-ubiquitous chain; tubulin B-alpha-1; Tubulin H2-alpha; tubulin K-alpha-1; tubulin(alpha1); tubulin, alpha 1; tubulin, alpha 1 (testis specific); tubulin, alpha 1, like; tubulin, alpha 1, like 2; tubulin, alpha 1a; tubulin, alpha 1b; tubulin, alpha 1c; tubulin, alpha 2; tubulin, alpha 3c; tubulin, alpha 3e; tubulin, alpha 4; tubulin, alpha 4a; tubulin, alpha 5; tubulin, alpha 6; tubulin, alpha, brain-specific; tubulin, alpha, ubiquitous; tubulin1alpha; tubulin84B; Tubulin-alpha1; Unknown (protein for MGC:133894); unnamed protein product; wu:fb22g06; wu:fb37a10; XELAEV_18019849mg; XELAEV_18020901mg; YML085C
Gene Symbols Tuba1a, Tuba1b, TUBA1C, TUBA3C, TUBA3E, TUBA4A, TUBAL3, TUBB2B
Host Species Rabbit
Immunogen A peptide corresponding to the C-terminus of alpha-Tubulin.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10376, 112714, 22142, 22143, 22145, 22146, 238463, 291081, 291287, 300218, 316531, 347733, 500929, 64158, 7277, 7278, 73710, 7846, 79861, 84790
Target Species Human, Mouse, Rat, All species
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.