Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha2 Adaptin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982108
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2108 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2108 Supplier Bioss Supplier No. BS1228R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adaptins are components of the adaptor complexes which link clathrin to receptors in coated vesicles. Clathrin-associated protein complexes are believed to interact with the cytoplasmic tails of membrane proteins, leading to their selection and concentration. Alpha adaptin is a subunit of the plasma membrane adaptor. It binds polyphosphoinositides.

Specifications

Antigen alpha2 Adaptin
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene AP2A2
Gene Accession No. O94973
Gene Alias 100 kDa coated vesicle protein C; 2410074K14Rik; adapter-related protein complex 2 alpha-2 subunit; adapter-related protein complex 2 subunit alpha-2; adaptin alpha A; Adaptin alpha B; adaptin, alpha B; Adaptor protein complex AP-2 subunit alpha-2; adaptor protein complex AP-2, alpha 2 subunit; adaptor related protein complex 2 alpha 2 subunit; adaptor related protein complex 2, alpha 2 subunit; Adaptor-related protein complex 2 subunit alpha-2; adaptor-related protein complex 2, alpha 2 subunit; adaptor-related protein complex AP-2, alpha 2 subunit; ADTAA; Adtab; AF006990; Alpha adaptin A; Alpha adaptin C; Alpha1 adaptin; Alpha2 adaptin; Alpha2-adaptin; Alpha-adaptin C; alpha-adaptin C; Huntingtin interacting protein J; alpha-C adaptin; AP2 alpha; AP-2 complex subunit alpha-2; AP2A1; Ap2a2; AW146353; C78001; CLAPA1; CLAPA2; Clathrin assembly; Clathrin assembly protein complex 2 alpha-C large chain; clathrin-associated/assembly/adaptor protein, large, alpha 2; HIP 9; HIP9; HIP-9; Huntingtin yeast partner J; Huntingtin-interacting protein 9; Huntingtin-interacting protein J; HYPJ; KIAA0899; L25; Plasma membrane adaptor HA2/AP2 adaptin alpha C subunit
Gene Symbols AP2A2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human AP2A2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.