Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AlphaA Crystallin/CRYAA Antibody (1H3.B8), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212875AF700
Description
AlphaA Crystallin/CRYAA Monoclonal specifically detects AlphaA Crystallin/CRYAA in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
AlphaA Crystallin/CRYAA | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CRYAA | |
Native Alpha Crystallin | |
200 μL | |
Primary | |
Detects approx 20kDa. Does not cross-react with aB-crystallin, Beta L-crystallin, BH- crystallin, gamma-crystallin, HSP25, HSP27 or HSP47 proteins. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
1H3.B8 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
CRYA1crystallin, alpha-1, crystallin, alpha A, Heat shock protein beta-4, HSPB4alpha-crystallin A chain, human alphaA-crystallin (CRYA1), 10HspB4 | |
Mouse | |
Protein G purified | |
Membrane Trafficking and Chaperones, Vision | |
1409 | |
Human, Mouse, Rat, Bovine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction