Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AlphaA Crystallin/CRYAA Antibody (1H3.B8), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP21287550UG
Description
AlphaA Crystallin/CRYAA Monoclonal specifically detects AlphaA Crystallin/CRYAA in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
AlphaA Crystallin/CRYAA | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence, Immunoblotting | |
CRYA1crystallin, alpha-1, crystallin, alpha A, Heat shock protein beta-4, HSPB4alpha-crystallin A chain, human alphaA-crystallin (CRYA1), 10HspB4 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Detects approx 20kDa. Does not cross-react with aB-crystallin, Beta L-crystallin, BH- crystallin, gamma-crystallin, HSP25, HSP27 or HSP47 proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoblot | |
1H3.B8 | |
Unconjugated | |
NP_000385.1 | |
CRYAA | |
Native Alpha Crystallin | |
50 μg | |
Membrane Trafficking and Chaperones, Vision | |
1409 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction