Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AlphaB Crystallin/CRYAB Antibody (CL10758), Novus Biologicals™

Mouse Monoclonal Antibody
$322.50 - $547.00
Specifications
Antigen | AlphaB Crystallin/CRYAB |
---|---|
Clone | CL10758 |
Dilution | Western Blot 1 ug/ml, Immunohistochemistry 1:20000 - 1:50000, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunohistochemistry-Paraffin 1:20000 - 1:50000 |
Applications | Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry |
Classification | Monoclonal |
Description
AlphaB Crystallin/CRYAB Monoclonal antibody specifically detects AlphaB Crystallin/CRYAB in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
AlphaB Crystallin/CRYAB | |
Western Blot 1 ug/ml, Immunohistochemistry 1:20000 - 1:50000, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunohistochemistry-Paraffin 1:20000 - 1:50000 | |
Monoclonal | |
Purified | |
RUO | |
Rat, Human | |
alpha B crystallin, alpha(B)-crystallin, alpha-crystallin B chain, CRYA2alpha crystallin B chain, crystallin, alpha B, CTPP2, Heat shock protein beta-5, heat-shock 20 kD like-protein, HSPB5, Renal carcinoma antigen NY-REN-27, Rosenthal fiber component | |
This antibody was developed against a Synthetic Peptide of P02511. Actual immunogen sequence is proprietary. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
CL10758 | |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Neurotransmission, Phospho Specific, Protein Phosphatase, Sensory Systems, Signal Transduction, Tyrosine Kinases, Vision | |
PBS, pH 7.2, 40% glycerol | |
1410 | |
IgG1 | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title