Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AlphaB Crystallin/CRYAB Antibody (CPTC-CRYAB-1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308380
Description
AlphaB Crystallin/CRYAB Monoclonal specifically detects AlphaB Crystallin/CRYAB in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.Specifications
AlphaB Crystallin/CRYAB | |
Monoclonal | |
1 mg/mL | |
Western Blot : 1-2 ug/mL, Immunohistochemistry-Paraffin : 1-2 ug/mL, Protein Array | |
alpha B crystallin, alpha(B)-crystallin, alpha-crystallin B chain, CRYA2alpha crystallin B chain, crystallin, alpha B, CTPP2, Heat shock protein beta-5, heat-shock 20 kD like-protein, HSPB5, Renal carcinoma antigen NY-REN-27, Rosenthal fiber component | |
Recombinant human full-length AlphaB Crystallin/CRYAB protein | |
100 μg | |
Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Neurotransmission, Phospho Specific, Protein Phosphatase, Sensory Systems, Signal Transduction, Tyrosine Kinases, Vision | |
1410 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG2c κ |
Western Blot, Immunohistochemistry (Paraffin), Peptide Array | |
CPTC-CRYAB-1 | |
Unconjugated | |
10mM PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction