Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AlphaB Crystallin/CRYAB Antibody (CRYAB/7918), Novus Biologicals™

Mouse Monoclonal Antibody
$216.50 - $443.50
Specifications
Antigen | AlphaB Crystallin/CRYAB |
---|---|
Clone | CRYAB/7918 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Description
AlphaB Crystallin/CRYAB Monoclonal antibody specifically detects AlphaB Crystallin/CRYAB in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
AlphaB Crystallin/CRYAB | |
0.2 mg/ml | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Neurotransmission, Phospho Specific, Protein Phosphatase, Sensory Systems, Signal Transduction, Tyrosine Kinases, Vision | |
10mM PBS with 0.05% BSA | |
1410 | |
IgG | |
Protein A or G purified |
CRYAB/7918 | |
Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human | |
alpha B crystallin, alpha(B)-crystallin, alpha-crystallin B chain, CRYA2alpha crystallin B chain, crystallin, alpha B, CTPP2, Heat shock protein beta-5, heat-shock 20 kD like-protein, HSPB5, Renal carcinoma antigen NY-REN-27, Rosenthal fiber component | |
Recombinant human full-length AlphaB Crystallin/CRYAB protein | |
Primary | |
Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title