Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AlphaProTech Cleanroom MVT Face Masks
BIOPHARMA

Catalog No. p-4829954 Shop All AlphaProTech Products
Click to view available options
Size:
Standard
This item is not returnable. View return policy
This item is not returnable. View return policy

Face masks with earloops are designed to shield mouth while allowing ease of breathing.

  • Clean mask secured with soft knit earloops
  • Disposable
  • Soft
  • White

Specifications

Product Type Cleanroom Face Mask
Color White
Size Standard
Sterility Non-sterile
Style Pleated
Disposable Single-use
For Use With (Application) Semiconductor, Disk Drive, Medical Device, Pharmaceutical, Biomedical
Model 9060
Packaging Type Double-bagged, Cleanroom Packaged
Series MVT
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.