Learn More
AlphaProTech Critical Cover™ Combo™ Face Masks with Shield
Catalog No. 191694066 Shop All AlphaProTech Products
Click to view available options
Fastening Type:
Earloops
This item is not returnable. View return policy
This item is not returnable. View return policy
Face mask with 7 mil thick eye shield features anti-fog coating and snaps for easy removal.
  • Fluid resistant mask
  • Shield attaches to mask with snaps
  • 7 mil optically clear shield with anti-fog shield coating, low glare and is distortion free
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 3
Color Blue
Fastening Type Earloops
Size Standard
Sterility Non-sterile
Style Pleated with Eyeshield
Disposable Single-use
Filter Efficiency ≥98% (PFE), ≥98% (BFE)
Includes Anti-fog coating optically clear full wrap around shield (7 mil thickness) with foam anti-splash forehead band
Model 5194-E
Pore Size 0.1 μm
Resistance Fluid
Series Critical Cover™ Combo™
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.