Learn More
Ansell AlphaTec™ 04-412 PVC-Nitrile Blend Coated Gauntlet Gloves
Catalog No. 19014903 Shop All Ansell Products
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-014903 Large
19-014904 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-014903 Supplier Ansell Supplier No. 211134
This item is not returnable. View return policy
Combination PVC and nitrile gloves feature a secure wet grip while providing chemical, cut and abrasion resistance.
Ideal for applications involving petrochemicals and for handling equipment and abrasive materials coated with acids, caustics, solvents, grease, and oil.
  • Feature PVC-nitrile blend rough finish
  • Curved, preflexed fingers and wing thumb give extra comfort and flexibility
  • Two-piece lining eliminates seams from work areas to prevent irritation and premature wear-through
TRUSTED_SUSTAINABILITY
Specifications
Color Green
Latex Free Yes
Material PVC, Nitrile
Size Large
For Use With (Application) Chemical processing, oil refining and petrochemicals, cleaning and de-greasing with soaps and detergent formulations, handling equipment and abrasive materials coated with oil, grease, acids and solvents.
Glove Cuff Style Gauntlet
Liner Material Jersey Knit
Lining Lined
Model 04-412 9 211134
Length (English) 12 in.
Length (Metric) 30.5 cm