Learn More
Description
- Liquid and chemical protection gloves
- Incorporates revolutionary Ansell Grip Technology™ to enable users to handle wet or oily objects with less grip force and more control
- Unique combination of liquid-proof chemical resistance and grip, together with flexibility and dexterity, makes the logical choice when working with chemicals
- Reliable chemical protection for added user safety
- Patented Ansell Grip Technology™ ensures that the coating polymer does not penetrate the liner during manufacture
- Coating is therefore consistent and sound, significantly reducing the likelihood of chemical leakage onto the skin
- AlphaTec™ models with white nylon liner are recommended for indoor applications, whereas with black acrylic liner are recommended for outdoor applications
Applications:
- Applying sealers, touching up
- Body painting including surface inspection
- Changing tools and dies
- Handling application and cleaning tools
- Loading and unloading process equipment
- Metalworking and assembly operations
- Opening furnaces draining pumps valves or lines and crackers BTX process
- Painting
- Painting or cleaning tools or robots
- Transferring liquids and solids between vessels and tanks and process equipment
- Warehousing of raw materials
Specifications
Specifications
| Certifications/Compliance | FDA 21 CFR 177-2600 |
| Applications | Industrial |
| Color | Black, Red |
| Latex Free | Yes |
| Material | Nitrile |
| Product Type | Chemical-resistant Gloves |
| Size | 9 |
| Thickness (English) | 13 mil |
| For Use With (Application) | Chemical plants and transportation, chemical processing, oil refining and petrochemicals, cleaning and de-greasing, exposure to chemicals, solvents, hazardous liquids, PVC cements or standing water, janitorial and maintenance work |
| Glove Cuff Style | Gauntlet |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.