Learn More
Ansell™ AlphaTec™ 66-667 Polyester Coverall
Catalog No. 19501051 Shop All Ansell Products
Change view
Click to view available options
Size:
Large
3X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-501-051 3X-Large
19-501-052 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-501-051 Supplier Ansell™ Supplier No. 666673INL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Breathable, chemical splash resistance, re-usable Level B protection
  • Chemical splash fabric provides protection with penetration data for more than 70 chemicals
  • Breathable fabric maximizes comfort, reducing the risk of heat stress and increasing productivity
  • Stitched and heat sealed seams and double front storm flap to protect from chemical spray
  • Machine washable
  • Made of a unique breathable membrane from Gore helping manage heat stress, while offering liquid chemical splash protection
  • Design Features:
    • Raglan sleeves, designed for comfortable arm movement
    • Adjustable hook and loop closure straps at the wrist, ankle and front flap for a snug fit
    • Double front flap for secured closure
    • Front tab to clip a microphone or other devices
    • Optional detachable hood (66-664) attached to collar by Velcro hook and loop
  • Uses:
    • Chemical, Food processing, Mining
  • TRUSTED_SUSTAINABILITY
    Specifications
    Size 3X-Large
    Material Polyester with GORE Membrane
    Color Red
    Seam Type Sealed
    Certifications/Compliance NFPA 1992 (Ed 2012)
    Weight (English) 2.87 lb.
    Weight (Metric) 1300 g
    Quantity 5 Cs.
    Adhere to
    Safety Regulations and Best Practices
    Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.